Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036325.3 | NP_001031402.1 | 7 - 8 | 932 | 131 | |
NM_001084478.1 | NP_001077947.1 | 7 - 8 | 852 | 131 | ||
NM_001335832.1 | NP_001318270.1 | 7 - 8 | 858 | 131 | ||
NM_001335833.1 | NP_001324656.1 | 6 - 7 | 983 | 131 | ||
NM_127863.5 | NP_179881.3 | 7 - 8 | 970 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |