Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335834.1 | NP_001324624.1 | 11 - 12 | 1189 | 102 | |
NM_127865.3 | NP_179883.1 | 11 - 12 | 1143 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Arabidopsis thaliana Probe spans exons |