Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127866.3 | NP_179884.1 | 9 - 10 | 966 | 74 | |
NM_201788.2 | NP_973517.1 | 9 - 10 | 912 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Arabidopsis thaliana Probe spans exons |