Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127902.6 | NP_179919.1 | 1 - 2 | 542 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |