Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335847.1 | NP_001324816.1 | 4 - 5 | 1013 | 106 | |
NM_127902.6 | NP_179919.1 | 9 - 10 | 2138 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |