Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198511.1 | NP_001185440.1 | 10 - 11 | 1141 | 73 | |
NM_106631.5 | NP_178100.3 | 10 - 11 | 1333 | 73 | ||
NM_180652.3 | NP_850983.1 | 10 - 11 | 1127 | 73 | ||
GenBank mRNA | AK317667.1 | - | 9 - 10 | 1054 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |