Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036226.2 | NP_001031303.1 | 11 - 12 | 1403 | 69 | |
NM_001198511.1 | NP_001185440.1 | 12 - 13 | 1304 | 69 | ||
NM_001334924.1 | NP_001323193.1 | 7 - 8 | 885 | 69 | ||
NM_106631.5 | NP_178100.3 | 12 - 13 | 1496 | 69 | ||
NM_180652.3 | NP_850983.1 | 12 - 13 | 1290 | 69 | ||
GenBank mRNA | AK317667.1 | - | 11 - 12 | 1217 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |