Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036226.2 | NP_001031303.1 | 4 - 5 | 647 | 60 | |
NM_001198511.1 | NP_001185440.1 | 4 - 5 | 404 | 60 | ||
NM_106631.5 | NP_178100.3 | 4 - 5 | 596 | 60 | ||
NM_180652.3 | NP_850983.1 | 4 - 5 | 390 | 60 | ||
GenBank mRNA | AK317667.1 | - | 3 - 4 | 317 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |