Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198511.1 | NP_001185440.1 | 9 - 10 | 997 | 101 | |
NM_106631.5 | NP_178100.3 | 9 - 10 | 1189 | 101 | ||
NM_180652.3 | NP_850983.1 | 9 - 10 | 983 | 101 | ||
GenBank mRNA | AK317667.1 | - | 8 - 9 | 910 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |