Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNEE/G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202581.2 | NP_001189510.1 | 12 - 13 | 2846 | 86 | |
NM_001335223.1 | NP_001318196.1 | 12 - 13 | 2877 | 86 | ||
NM_001335224.1 | NP_001325371.1 | 12 - 13 | 2843 | 86 | ||
NM_001335225.1 | NP_001325372.1 | 11 - 12 | 3025 | 86 | ||
NM_001335226.1 | NP_001325370.1 | 11 - 12 | 3022 | 86 | ||
NM_126460.4 | NP_178508.2 | 9 - 10 | 2111 | 86 | ||
NM_180656.2 | NP_850987.1 | 12 - 13 | 2923 | 86 | ||
NM_180657.2 | NP_850988.1 | 12 - 13 | 2768 | 86 | ||
GenBank mRNA | AF450479.1 | - | 12 - 13 | 2923 | 86 | |
AJ508604.1 | - | 10 - 11 | 2251 | 86 | ||
AY093213.1 | - | 9 - 10 | 2111 | 86 | ||
BT008793.1 | - | 8 - 9 | 1792 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |