Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNEE/G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202581.2 | NP_001189510.1 | 5 - 6 | 1903 | 85 | |
NM_001335223.1 | NP_001318196.1 | 5 - 6 | 1934 | 85 | ||
NM_001335224.1 | NP_001325371.1 | 5 - 6 | 1900 | 85 | ||
NM_001335225.1 | NP_001325372.1 | 4 - 5 | 2082 | 85 | ||
NM_001335226.1 | NP_001325370.1 | 4 - 5 | 2079 | 85 | ||
NM_126460.4 | NP_178508.2 | 2 - 3 | 1168 | 85 | ||
NM_180656.2 | NP_850987.1 | 5 - 6 | 1980 | 85 | ||
NM_180657.2 | NP_850988.1 | 5 - 6 | 1825 | 85 | ||
NM_180658.2 | NP_850989.1 | 2 - 3 | 1168 | 85 | ||
GenBank mRNA | AF450479.1 | - | 5 - 6 | 1980 | 85 | |
AJ508604.1 | - | 3 - 4 | 1308 | 85 | ||
AY093213.1 | - | 2 - 3 | 1168 | 85 | ||
BT008793.1 | - | 1 - 2 | 849 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |