Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126703.4 | NP_178744.3 | 4 - 5 | 353 | 90 | |
NM_180659.3 | NP_850990.1 | 4 - 5 | 392 | 90 | ||
GenBank mRNA | AY048212.1 | - | 4 - 5 | 305 | 90 | |
AY087310.1 | - | 4 - 5 | 356 | 90 | ||
AY094019.1 | - | 4 - 5 | 271 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |