Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337803.1 | NP_001326171.1 | 6 - 7 | 890 | 88 | |
NM_111750.4 | NP_566344.2 | 6 - 7 | 905 | 88 | ||
NM_180665.1 | NP_850996.1 | 6 - 7 | 830 | 88 | ||
NM_202535.1 | NP_974264.1 | 5 - 6 | 1006 | 88 | ||
GenBank mRNA | AB045112.1 | - | 6 - 7 | 863 | 88 | |
AY099706.1 | - | 6 - 7 | 906 | 88 | ||
AY128900.1 | - | 6 - 7 | 768 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |