Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339361.1 | NP_001326364.1 | 1 - 2 | 623 | 60 | |
NM_001339362.1 | NP_001326363.1 | 2 - 3 | 456 | 60 | ||
NM_148859.4 | NP_680112.2 | 1 - 2 | 278 | 60 | ||
NM_180682.4 | NP_851013.2 | 2 - 3 | 435 | 60 | ||
GenBank mRNA | BT029372.1 | - | 1 - 2 | 278 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |