Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036800.2 | NP_001031877.1 | 12 - 13 | 1174 | 79 | |
NM_001343292.1 | NP_001318556.1 | 12 - 13 | 1174 | 79 | ||
NM_121378.5 | NP_568290.3 | 10 - 11 | 965 | 79 | ||
NM_180705.3 | NP_851036.1 | 12 - 13 | 1570 | 79 | ||
GenBank mRNA | AY039542.1 | - | 12 - 13 | 1119 | 79 | |
AY062867.1 | - | 12 - 13 | 1164 | 79 | ||
AY102150.1 | - | 12 - 13 | 1056 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |