Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121378.5 | NP_568290.3 | 12 - 13 | 1118 | 99 | |
NM_180705.3 | NP_851036.1 | 14 - 15 | 1723 | 99 | ||
GenBank mRNA | AY039542.1 | - | 14 - 15 | 1272 | 99 | |
AY062867.1 | - | 14 - 15 | 1317 | 99 | ||
AY102150.1 | - | 14 - 15 | 1209 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |