Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036800.2 | NP_001031877.1 | 9 - 10 | 815 | 134 | |
NM_001343292.1 | NP_001318556.1 | 9 - 10 | 815 | 134 | ||
NM_121378.5 | NP_568290.3 | 7 - 8 | 606 | 134 | ||
NM_180705.3 | NP_851036.1 | 9 - 10 | 1211 | 134 | ||
GenBank mRNA | AY039542.1 | - | 9 - 10 | 760 | 134 | |
AY062867.1 | - | 9 - 10 | 805 | 134 | ||
AY102150.1 | - | 9 - 10 | 697 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |