Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203415.1 | NP_001190344.1 | 10 - 11 | 1523 | 106 | |
NM_180712.3 | NP_851043.1 | 10 - 11 | 1485 | 106 | ||
GenBank mRNA | AF360208.1 | - | 10 - 11 | 1410 | 106 | |
AY040056.1 | - | 10 - 11 | 1295 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |