Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203418.1 | NP_001190347.1 | 1 - 2 | 238 | 61 | |
GenBank mRNA | AK227214.1 | - | 1 - 2 | 221 | 61 | |
AK318923.1 | - | 1 - 2 | 222 | 61 | ||
AY090237.1 | - | 1 - 2 | 224 | 61 | ||
BT000632.1 | - | 1 - 2 | 167 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |