Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHF2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122125.3 | NP_568410.1 | 3 - 4 | 439 | 96 | |
NM_180719.4 | NP_851050.1 | 3 - 4 | 440 | 96 | ||
NM_180720.1 | NP_851051.1 | 3 - 4 | 300 | 96 | ||
GenBank mRNA | AF079182.1 | - | 3 - 4 | 351 | 96 | |
AF324994.1 | - | 3 - 4 | 317 | 96 | ||
BT002347.1 | - | 2 - 3 | 186 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |