Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHF2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343714.1 | NP_001331677.1 | 4 - 5 | 701 | 80 | |
NM_122125.3 | NP_568410.1 | 6 - 7 | 966 | 80 | ||
NM_180719.4 | NP_851050.1 | 6 - 7 | 967 | 80 | ||
NM_180720.1 | NP_851051.1 | 6 - 7 | 827 | 80 | ||
GenBank mRNA | AF079182.1 | - | 6 - 7 | 878 | 80 | |
AF324994.1 | - | 6 - 7 | 844 | 80 | ||
BT002347.1 | - | 5 - 6 | 713 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |