Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343718.1 | NP_001318617.1 | 6 - 7 | 974 | 99 | |
NM_122126.5 | NP_568411.4 | 8 - 9 | 1100 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |