Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343718.1 | NP_001318617.1 | 12 - 13 | 2502 | 80 | |
NM_122126.5 | NP_568411.4 | 14 - 15 | 2628 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |