Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085146.2 | NP_001078615.1 | 2 - 3 | 642 | 87 | |
NM_001085147.2 | NP_001078616.1 | 2 - 3 | 143 | 87 | ||
NM_001343793.1 | NP_001331558.1 | 1 - 2 | 756 | 87 | ||
NM_122227.2 | NP_197712.2 | 2 - 3 | 143 | 87 | ||
NM_180731.3 | NP_851062.2 | 2 - 3 | 663 | 87 | ||
GenBank mRNA | AK228998.1 | - | 2 - 3 | 143 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |