Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GUS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122885.2 | NP_198344.2 | 1 - 2 | 150 | ||
NM_180762.3 | NP_851093.1 | 1 - 2 | 392 | 150 | ||
GenBank mRNA | AK229275.1 | - | 1 - 2 | 252 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |