Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344299.1 | NP_001318708.1 | 1 - 2 | 428 | 76 | |
NM_123328.2 | NP_568567.1 | 1 - 2 | 428 | 76 | ||
NM_180775.3 | NP_851106.1 | 1 - 2 | 456 | 76 | ||
GenBank mRNA | BT015791.1 | - | 1 - 2 | 234 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |