Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344353.1 | NP_001318716.1 | 5 - 6 | 1443 | 69 | |
NM_001344354.1 | NP_001332682.1 | 5 - 6 | 1255 | 69 | ||
NM_180778.4 | NP_851109.1 | 5 - 6 | 1339 | 69 | ||
GenBank mRNA | AF367281.1 | - | 5 - 6 | 1186 | 69 | |
AY133550.1 | - | 5 - 6 | 1083 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |