Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123861.3 | NP_199306.1 | 3 - 4 | 1419 | 94 | |
NM_180801.2 | NP_851132.1 | 3 - 4 | 1419 | 94 | ||
GenBank mRNA | AY048259.1 | - | 3 - 4 | 1407 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |