Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344582.1 | NP_001332492.1 | 5 - 6 | 3485 | 74 | |
NM_001344583.1 | NP_001332491.1 | 4 - 5 | 3191 | 74 | ||
NM_123873.3 | NP_199318.2 | 6 - 7 | 3401 | 74 | ||
NM_180802.2 | NP_851133.1 | 5 - 6 | 3653 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |