Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124156.5 | NP_199593.2 | 10 - 11 | 1800 | 101 | |
NM_180820.2 | NP_851151.1 | 10 - 11 | 1821 | 101 | ||
GenBank mRNA | AY158083.1 | - | 10 - 11 | 1471 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |