Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125941.1 | NP_001119413.1 | 12 - 13 | 1386 | 68 | |
NM_124502.3 | NP_199936.2 | 13 - 14 | 1378 | 68 | ||
NM_180837.3 | NP_851168.1 | 13 - 14 | 1470 | 68 | ||
GenBank mRNA | AB053171.1 | - | 13 - 14 | 1315 | 68 | |
AY140086.1 | - | 13 - 14 | 1327 | 68 | ||
BT008388.1 | - | 12 - 13 | 1067 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |