Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT5G51630 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203587.1 | NP_001190516.1 | 2 - 3 | 778 | 66 | |
NM_001344946.1 | NP_001330479.1 | 2 - 3 | 1015 | 66 | ||
NM_001344947.1 | NP_001330480.1 | 2 - 3 | 821 | 66 | ||
NM_001344948.1 | NP_001330481.1 | 2 - 3 | 756 | 66 | ||
NM_124542.3 | NP_199976.2 | 2 - 3 | 960 | 66 | ||
NM_180841.3 | NP_851172.1 | 1 - 2 | 1350 | 66 | ||
GenBank mRNA | AK228948.1 | - | 1 - 2 | 503 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |