Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT5G51630 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203587.1 | NP_001190516.1 | 4 - 5 | 1727 | 103 | |
NM_001344946.1 | NP_001330479.1 | 3 - 4 | 2108 | 103 | ||
NM_001344947.1 | NP_001330480.1 | 3 - 4 | 1914 | 103 | ||
NM_001344948.1 | NP_001330481.1 | 3 - 4 | 1849 | 103 | ||
NM_124542.3 | NP_199976.2 | 3 - 4 | 2053 | 103 | ||
NM_180841.3 | NP_851172.1 | 2 - 3 | 2443 | 103 | ||
GenBank mRNA | AK228948.1 | - | 2 - 3 | 1596 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |