Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124600.3 | NP_200034.1 | 2 - 3 | 329 | 145 | |
NM_180844.2 | NP_851175.1 | 2 - 3 | 211 | 145 | ||
GenBank mRNA | AK227533.1 | - | 2 - 3 | 193 | 145 | |
BT004590.1 | - | 1 - 2 | 94 | 145 | ||
U46924.1 | - | 2 - 3 | 300 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |