Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124600.3 | NP_200034.1 | 4 - 5 | 579 | 69 | |
NM_180844.2 | NP_851175.1 | 4 - 5 | 461 | 69 | ||
GenBank mRNA | AK227533.1 | - | 4 - 5 | 443 | 69 | |
BT004590.1 | - | 3 - 4 | 344 | 69 | ||
U46924.1 | - | 4 - 5 | 550 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |