Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085279.1 | NP_001078748.1 | 9 - 10 | 875 | 114 | |
NM_001085280.1 | NP_001078749.1 | 8 - 9 | 780 | 114 | ||
NM_001203598.1 | NP_001190527.1 | 8 - 9 | 1130 | 114 | ||
NM_001345033.1 | NP_001332707.1 | 8 - 9 | 906 | 114 | ||
NM_124691.3 | NP_568785.1 | 9 - 10 | 912 | 114 | ||
NM_180847.2 | NP_851178.1 | 8 - 9 | 972 | 114 | ||
NM_180848.3 | NP_851179.1 | 8 - 9 | 959 | 114 | ||
GenBank mRNA | AY040013.1 | - | 8 - 9 | 938 | 114 | |
AY079367.1 | - | 7 - 8 | 694 | 114 | ||
BT000742.1 | - | 8 - 9 | 923 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |