Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HGO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124787.4 | NP_200219.1 | 8 - 9 | 1216 | 78 | |
NM_180856.4 | NP_851187.1 | 8 - 9 | 1216 | 78 | ||
GenBank mRNA | AX417711.1 | - | 8 - 9 | 1161 | 78 | |
AY140075.1 | - | 8 - 9 | 1192 | 78 | ||
BT010329.1 | - | 8 - 9 | 1161 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |