Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345242.1 | NP_001331281.1 | 2 - 3 | 376 | 63 | |
NM_125093.6 | NP_200521.3 | 2 - 3 | 425 | 63 | ||
NM_180869.3 | NP_851200.1 | 2 - 3 | 267 | 63 | ||
GenBank mRNA | AJ249352.1 | - | 2 - 3 | 269 | 63 | |
AK226961.1 | - | 1 - 2 | 191 | 63 | ||
AY069869.1 | - | 2 - 3 | 269 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |