Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHOT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345288.1 | NP_001318824.1 | 16 - 17 | 2583 | 110 | |
NM_001345289.1 | NP_001330355.1 | 15 - 16 | 2222 | 110 | ||
NM_001345290.1 | NP_001330356.1 | 14 - 15 | 2572 | 110 | ||
NM_125199.2 | NP_568874.2 | 15 - 16 | 2042 | 110 | ||
NM_180879.2 | NP_851210.1 | 15 - 16 | 2399 | 110 | ||
NM_180880.2 | NP_851211.1 | 15 - 16 | 2395 | 110 | ||
NM_180881.3 | NP_851212.2 | 14 - 15 | 2180 | 110 | ||
GenBank mRNA | AF053941.2 | - | 14 - 15 | 1937 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |