Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHOT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345288.1 | NP_001318824.1 | 19 - 20 | 2851 | 99 | |
NM_001345289.1 | NP_001330355.1 | 18 - 19 | 2490 | 99 | ||
NM_001345290.1 | NP_001330356.1 | 17 - 18 | 2840 | 99 | ||
NM_125199.2 | NP_568874.2 | 17 - 18 | 2525 | 99 | ||
NM_180879.2 | NP_851210.1 | 18 - 19 | 2667 | 99 | ||
NM_180880.2 | NP_851211.1 | 18 - 19 | 2663 | 99 | ||
NM_180881.3 | NP_851212.2 | 17 - 18 | 2448 | 99 | ||
GenBank mRNA | AF053941.2 | - | 17 - 18 | 2205 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |