Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125995.2 | NP_001119467.1 | 3 - 4 | 654 | 140 | |
NM_001345427.1 | NP_001332136.1 | 3 - 4 | 845 | 140 | ||
NM_001345428.1 | NP_001332137.1 | 3 - 4 | 741 | 140 | ||
NM_125453.6 | NP_200868.2 | 3 - 4 | 573 | 140 | ||
NM_180902.5 | NP_851233.1 | 3 - 4 | 654 | 140 | ||
GenBank mRNA | AF434673.1 | - | 3 - 4 | 513 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |