Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XRCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345633.1 | NP_001330648.1 | 5 - 6 | 847 | 81 | |
NM_001345634.1 | NP_001330645.1 | 5 - 6 | 817 | 81 | ||
NM_001345635.1 | NP_001318876.1 | 5 - 6 | 847 | 81 | ||
NM_001345636.1 | NP_001330643.1 | 5 - 6 | 895 | 81 | ||
NM_001345637.1 | NP_001330642.1 | 5 - 6 | 858 | 81 | ||
NM_001345638.1 | NP_001330647.1 | 5 - 6 | 858 | 81 | ||
NM_001345639.1 | NP_001330646.1 | 5 - 6 | 858 | 81 | ||
NM_001345641.1 | NP_001330644.1 | 5 - 6 | 847 | 81 | ||
NM_125848.3 | NP_201257.2 | 5 - 6 | 858 | 81 | ||
NM_180937.2 | NP_851268.1 | 5 - 6 | 858 | 81 | ||
GenBank mRNA | AJ421043.1 | - | 5 - 6 | 777 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |