Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATH13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125893.5 | NP_201299.2 | 6 - 7 | 1257 | 97 | |
NM_180940.2 | NP_851271.1 | 6 - 7 | 1257 | 97 | ||
GenBank mRNA | AY059877.1 | - | 6 - 7 | 1240 | 97 | |
AY064044.1 | - | 6 - 7 | 1216 | 97 | ||
AY096382.1 | - | 6 - 7 | 1095 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |