Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATH13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125893.5 | NP_201299.2 | 7 - 8 | 1366 | 117 | |
NM_180940.2 | NP_851271.1 | 7 - 8 | 1366 | 117 | ||
GenBank mRNA | AY059877.1 | - | 7 - 8 | 1349 | 117 | |
AY064044.1 | - | 7 - 8 | 1325 | 117 | ||
AY096382.1 | - | 7 - 8 | 1204 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |