Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125900.2 | NP_201306.2 | 11 - 12 | 1446 | 75 | |
NM_180941.3 | NP_851272.1 | 11 - 12 | 1601 | 75 | ||
GenBank mRNA | AF367340.1 | - | 11 - 12 | 1445 | 75 | |
AY124866.1 | - | 11 - 12 | 1412 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |