Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203692.1 | NP_001190621.1 | 1 - 2 | 704 | 85 | |
NM_125941.2 | NP_569012.2 | 1 - 2 | 570 | 85 | ||
NM_180943.2 | NP_851274.1 | 1 - 2 | 821 | 85 | ||
GenBank mRNA | AK228019.1 | - | 544 | 85 | ||
AY050996.1 | - | 1 - 2 | 550 | 85 | ||
AY079350.1 | - | 1 - 2 | 483 | 85 | ||
AY085088.1 | - | 1 - 2 | 570 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |