Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345703.1 | NP_001318885.1 | 3 - 4 | 999 | 71 | |
NM_180944.3 | NP_851275.1 | 3 - 4 | 1294 | 71 | ||
GenBank mRNA | AF334715.1 | - | 2 - 3 | 771 | 71 | |
U18411.1 | - | 3 - 4 | 1004 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |