Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180944.3 | NP_851275.1 | 5 - 6 | 1500 | 93 | |
GenBank mRNA | AF334715.1 | - | 4 - 5 | 977 | 93 | |
U18411.1 | - | 5 - 6 | 1210 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |