Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115043.3 | NP_190752.1 | 7 - 8 | 914 | 70 | |
GenBank mRNA | AB017643.1 | - | 7 - 8 | 620 | 70 | |
AY087793.1 | - | 7 - 8 | 639 | 70 | ||
AY094441.1 | - | 7 - 8 | 736 | 70 | ||
AY125536.1 | - | 7 - 8 | 581 | 70 | ||
U72505.1 | - | 7 - 8 | 606 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |