Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chx28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115067.3 | NP_190776.2 | 3 - 4 | 1910 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |